missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MSA 2
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
MSA 2 is a non-nucleotide STING agonist (EC50 values are 8.3 and 24 μM for human STING isoforms WT and HAQ, respectively). Exhibits antitumor activity and stimulates interferon-β secretion in tumors. Induces tumor regression with durable antitumor immunity, and synergizes with anti-PD-1 in syngeneic mouse tumor models. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 4-(5,6-Dimethoxybenzo[b]thiophen-2-yl)-4-oxobutanoic acid |
| CAS | 129425-81-6 |
| Quantity | 10 mg |
| Target | STING Agonists |
| Molecular Formula | C14H14O5S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction