missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ MSCGM™ Mesenchymal Stem Cell Growth Medium BulletKit™

Product Code. 11620361
Change view
Click to view available options
Quantity:
440 mL Medium, 50 mL FBS, 10 mL L-Glutamine, 0.5 mL GA-1000
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11620361 440 mL Medium, 50 mL FBS, 10 mL L-Glutamine, 0.5 mL GA-1000 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11620361 Supplier Lonza™ Supplier No. PT3001
  • ONLINE EXCLUSIVE PRICE
    620.00 €
    517.08 € / Pack of 1
    Save 102.92 €
    16% Off
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Serum-containing medium designed to proliferate human bone marrow derived mesenchymal cells in an undifferentiated state.

  • Serum-containing medium.
  • Designed to proliferate human bone marrow derived mesenchymal cells in an undifferentiated state.
  • Screened to ensure that the serum neither promotes spontaneous differentiation of cells nor inhibits cell differentiation to osteogenic, chondrogenic, or adipogenic lineages (when properly stimulated with an appropriate differentiation media).
  • Includes both the basal media and the necessary supplements for proliferation of human bone marrow derived mesenchymal stem cells.
    • The basal medium (MSCBM™hMSC Basal Medium; catalog no. PT-3238) and the necessary supplements (MSCGM™hMSC SingleQuot Kit; catalog no. PT-4105) can also be purchased individually.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mesenchymal Stem Cells
Product Type Stem Cell Media and Supplements
Quantity 440 mL Medium, 50 mL FBS, 10 mL L-Glutamine, 0.5 mL GA-1000
pH Medium: 7.0 to 7.7
Color Medium: Red
Content And Storage MSCBM™ Basal Media (PT-3238), GA-100, MSCGM™ SingleQuots Supplement Kit (PT-4105), L-Glutamine, Fetal bovine serum; Store in dark at 2 to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.