missing translation for 'onlineSavingsMsg'
Learn More

MSI-1701, MedChemExpress

Product Code. 16485028
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485028

Brand: Medchem Express HY12219B

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress MSI-1701 is an analogue of MSI-1436 which can control weight gain and blood glucose level extracted from patent US 7410959 B1.

MSI-1701 is an analogue of MSI-1436 which can control weight gain and blood glucose level extracted from patent US 7410959 B1.

Specifications

Chemical Name or Material MSI-1701
CAS 390808-64-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C37H72N4O5S
Quantity 10 mg
SMILES NCCCNCCCCNCCCN[C@H]1C[C@@](C[C@@H](O)[C@]2([H])[C@]3([H])CC[C@@]4(C)[C@@]2([H])CC[C@@]4([C@H](C)CC[C@@H](OS(=O)(O)=O)C(C)C)[H])([H])[C@]3(C)CC1
Molecular Weight (g/mol) 685.06
Formula Weight 685.06
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.