missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ MSTFA Silylation Reagent

Product Code. 10319827 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
Unit Size:
100mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10319827 100 mL 100mL
10535631 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10319827 Supplier Macherey-Nagel™ Supplier No. 701270.1100
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Complete silylation reactions with high reaction rates, even without a catalyst. MSTFA Silylation Reagent is the most volatile trimethylsilyl amide available.

  • N-methyl-N-trimethylsilyl-trifluoroacetamide
  • M 199.1g/mol, Bp 70°C (75 mm Hg), density d20°/4° = 1.11
  • MSTFA: R' = CF3, R″ = CH3
  • The most volatile trimethylsilyl amide available
  • Very strong TMS donor which does not cause any noticeable FID fouling even after long-time measuring series
  • The already good solution characteristics can be improved by addition of submolar quantities of protic solvents (e.g., TFA for extremely polar compounds such as hydrochlorides) or pyridine (e.g., for carbohydrates).
  • Recommended application: carboxylic acids, hydroxy and ketocarboxylic acids, amino acids, amines, alcohols, polyalcohols, sugars, mercaptans and similar compounds with active hydrogen atoms. Even amine hydrochlorides can be silylated directly.
  • Advantages: complete reaction with high reaction rates, even without a catalyst (1–2 % TMCS or TSIM)
  • The by-product of the reaction (N-methyltrifluoroacetamide) features high volatility and short retention time.

Specifications

Quantity 100 mL
compliance-icons compliance-icons
Product Identifier
  • MSTFA
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash with water thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.