missing translation for 'onlineSavingsMsg'
Learn More

MSTFA For Gas Chromatography, Chem Lab

Product Code. 15989286
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15989286 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15989286 Supplier Chem-Lab Supplier No. CL00.1375.0010
  • ONLINE EXCLUSIVE PRICE
    245.00 €
    118.00 € / 10mL
    Save 127.00 €
    51% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Quantity 10 mL
Physical Form Liquid
Color Yellow
Melting Point -10 °C
Boiling Point 47.5 °C
Flash Point 23 °C
Density 0.969 g/mL
Molecular Weight (g/mol) 257.4 g/mol
Packaging Glass bottles
Assay > 99%
CAS 24589-78-4
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • MSTFA
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: MSTFA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.