missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent

Product Code. 11786957 Shop All Thermo Scientific Products
Change view
Click to view available options
Standard Components:
MSTFA
MSTFA + 1% TMCS
Quantity:
10 g
10 x 1 mL ampules
100 mL
25 mL
Unit Size:
25mL
Each
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Components Quantity Unit Size
11786957 MSTFA 10 x 1 mL ampules Pack of 10
11587670 MSTFA 10 g Each
11507680 MSTFA 100 mL Each
11567851 MSTFA + 1% TMCS 10 x 1 mL ampules Pack of 10
11597670 MSTFA 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11786957 Supplier Thermo Scientific™ Supplier No. TS48910
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offer maximum volatility with the Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent.

Use Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent for GC analyses of early eluting compounds that would otherwise be obscured in the chromatogram. MSTFA, the most volatile TMS-amide available, and its byproduct N-methyltrifluoroacetamide typically elute with the solvent front, allowing TMS derivatives of small molecules to be analyzed. Addition of Thermo Scientific™ TMCS aids derivatization of amides, secondary amines, and hindered hydroxyls not derivatized by MSTFA alone.

  • Trimethylsilyl donor strength comparable to BSA and BSTFA
  • Reacts to replace labile hydrogens on a wide range of polar compounds with a Si(CH3)3 group
  • Used to prepare volatile and thermally stable derivatives for GC and MS
  • Volatile byproduct, N-methyltrifluoroacetamide, has an even lower retention time than MSTFA
  • Often TMS derivatives of small molecules can be analyzed when derivatized with MSTFA because the byproducts and the reagent itself usually elute with the solvent front
  • Addition of Thermo Scientfic TMCS aids derivatization of amides, secondary amines and hindered hydroxyls not derivatized by MSTFA alone
TRUSTED_SUSTAINABILITY

Specifications

For Use With GC/MS
Hazards Flammable, Irritant
Standard Concentration 98 to 100%
Storage 2°C to 8°C
Quantity 10 x 1 mL ampules
Standard Components MSTFA
compliance-icons compliance-icons
Product Identifier
  • N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P243-Take precautionary measures against static discharge.
  • P264-Wash thoroughly after handling.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Acetamide, 2, 2, 2-trifluoro-N-methyl-N-(trimethylsilyl)-

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.