missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- A new grinding/milling chamber extends the application range of the IKA TubeMill control
- Stainless steel beater is bended in a 45° angle
- Suitable for mixing of fine powders
- Bended beater creates a more effective flow in the chamber which leads to an even better grinding result
Specifications
Specifications
| Product Type | Grinding Chamber |
| Capacity (Metric) | 40 mL |
| For Use With (Equipment) | Tube Mill |
| Packaging Quantity | 100/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?