Learn More
IKA™ MT Disposable Grinding Chamber
Product Code. 15861782 Shop All IKA Products
Change view
Click to view available options
Packaging Quantity:
10/Pk.
50/Pk.
Unit Size:
Pack of 10
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity unitSize
15861782 50/Pk. Pack of 50
15761907 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15861782 Supplier IKA™ Supplier No. 0020006984
In Stock
Add to basket
Add to basket
Clear, disposable grinding chamber for milling, mixing, and grinding.
  • Disposable, one-time use
  • Clear chamber for easy viewing
TRUSTED_SUSTAINABILITY
Specifications
Product Type Disposable Grinding Chamber
Capacity (Metric) 100 mL
Packaging Quantity 50/Pk.