missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent

Product Code. 11597861 Shop All Thermo Scientific Products
Change view
Click to view available options
Standard Components:
MTBSTFA
MTBSTFA + 1% TBMDS
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Components Unit Size
11597861 MTBSTFA + 1% TBMDS Pack of 10
11527680 MTBSTFA Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11597861 Supplier Thermo Scientific™ Supplier No. TS48927
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Produce stable TBDMS derivatives for GC analysis with the Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent.

Produce stable TBDMS derivatives for GC analysis with the Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent. MTBSTFA derivatizes hydroxyl, carboxyl, thiol and primary and secondary amines with typically >96% yields, and the reaction byproducts are volatile and neutral. TBDMS ethers are 104 times more stable to hydrolysis than the corresponding TMS ethers. Addition of Thermo Scientific™ TBDMCS increases silylation potential.

Additional Product Information:
  •  Derivatives have a high molecular concentration at M-57.
TRUSTED_SUSTAINABILITY

Specifications

For Use With GC
Hazards Irritant
Standard Concentration 98 to 100, 0.1 to 1%
Storage 2°C to 8°C
Quantity 10 x 1 mL ampules
Standard Components MTBSTFA + 1% TBMDS

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.