missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies mTeSR™1 Without Phenol Red
Description
- mTeSR™1 is the most widely published feeder-free cell culture medium for human embryonic stem cells (ES cells) and induced pluripotent stem cells (iPS cells), with established protocols for applications ranging from derivation to differentiation
- It has been used to successfully maintain hundreds of ES and iPS cell lines in over 40 countries, and has supported top pluripotent stem cell publications and researchers
- mTeSR™1 is a highly specialized, serum-free, and complete cell culture medium
- With pre-screened raw materials that ensure batch-to-batch consistency and robust feeder-free protocols for ES and iPS cell culture, mTeSR™1 provides more consistent cultures with homogeneous, undifferentiated phenotypes
- mTeSR™1 is now available without phenol red, for applications in which the presence of phenol red is undesirable
Specifications
Specifications
| Cell Type | Pluripotent Stem Cells |
| For Use With (Application) | Cell Culture, Expansion, Maintenance |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?