missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MTK 458

Product Code. 18044812
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18044812 10 mg 10mg
18004822 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18044812 Supplier Tocris Bioscience™ Supplier No. 7980/10
  • ONLINE EXCLUSIVE PRICE
    179.00 €
    169.05 € / 10mg
    Save 9.95 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

PINK1 activator

MTK 458 is a brain penetrant PTEN-induced kinase 1 (PINK1) activator. It binds to and stabilizes the active form of PINK1, increasing its activity and activating mitophagy downstream. It mediates clearance of pathogenic α-synuclein and normalizes pS65-Ubiquitin (pUb) in in vivo Parkinson's models. MTK458 is orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(1R)-1,2,3,4-Tetrahydro-1-naphthalenyl]-6-(trifluoromethyl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine
CAS 2499962-58-0
Quantity 10 mg
Target PINK1
Molecular Formula C17H15F3N4
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.