missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MU 1700 trihydrochloride
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
MU 1700 trihydrochloride is a potent and selective ALK2 and ALK1 inhibitor (IC50 values are 6 nM and 13 nM, respectively in biochemical assays). It is also a potent ALK6 inhibitor (IC50 = 41 nM). It shows selectivity for ALK over >360 other kinases. It shows favorable pharmacokinetic for in vivo use. MU 1700 trihydrochloride is orally bioavailable. It is cell membrane permeable and brain penetrant.
Specifications
Specifications
| Chemical Name or Material | 6-(4-Piperazin-1-ylphenyl)-3-quinolin-4-ylfuro[3,2-b]pyridine trihydrochloride |
| Quantity | 10 mg |
| Target | ALK Inhibitors |
| Molecular Formula | C26H22N4O.3HCl |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction