missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Mucasol™ Universal Detergent Liquid Concentrate

Product Code. p-8000881 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
5 L
Container Type:
Jerrican
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Container Type unitSize
10739301 5 L Jerrican 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10739301 Supplier Brand™ Supplier No. 44005

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Cleans instruments made of glass, plastic, rubber, and metal in 10 to 30 minutes without mechanical help. BRAND™ Mucasol™ Universal Detergent Liquid Concentrate replaces chromosulfuric acid, is free from chlorine and caustic alkalis, is non-toxic, nonaggressive and biodegradable.

  • Liquid, alkaline high-performance concentrate for immersion and ultrasonic baths. Universal cleaner for laboratory equipment and instruments made of glass, porcelain, plastic, rubber and metal
  • Items are cleaned in about 10 to 30 minutes, without mechanical help
  • Mucasol™ replaces chromosulfuric acid, is free from chlorine and caustic alkalis, is non-toxic, nonaggressive and biodegradable
  • Prolonged exposure of zinc and non-anodized aluminum surfaces should be avoided
TRUSTED_SUSTAINABILITY

Specifications

Product Type Mucasol Universal Detergent
pH ∼11.5 (0.7%), ∼11.7 (3%)
Appearance Light Green
Composition Alkaline; >30% phosphates, <5% anionic surfactants, amphoteric surfactants, complexing agents,
Concentration Ultrasonic bath: 0.2 to 2% (1.4 to 14 mL/L); Immersion bath: 0.7 to 3% (5 to 22 mL/L)
Container Type Jerrican
For Use With (Application) For immersion and ultrasonic baths. Universal cleaner for laboratory equipment and instruments made of glass, porcelain, plastic, rubber and Metal
Quantity 1 Ea.
Volume (Metric) 5 L
compliance-icons
Product Identifier
  • mucasol
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Potassium PhosphateTribasic Trihydrate,Tetrapotassium pyrophosphate, Orthophosphoric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.