missing translation for 'onlineSavingsMsg'
Learn More

Mucic acid, 98%

Product Code. 11441197
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441197 100 g 100g
11451197 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441197 Supplier Thermo Scientific Alfa Aesar Supplier No. B24170.22
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 526-99-8 | C6H10O8 | 210.138 g/mol

Mucic acid is used to replace tartaric acid in self-rising flour or fizzies. It is also used as a precursor of adipic acid en route to nylon by a rhenium-catalyzed deoxydehydration reaction. It replaces potassium bitartrate in baking powder and also used for manufacture of granular effervescing salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mucic acid is used to replace tartaric acid in self-rising flour or fizzies. It is also used as a precursor of adipic acid en route to nylon by a rhenium-catalyzed deoxydehydration reaction. It replaces potassium bitartrate in baking powder and also used for manufacture of granular effervescing salts.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 526-99-8
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.138
MDL Number MFCD00004239
InChI Key DSLZVSRJTYRBFB-DUHBMQHGSA-N
Synonym galactaric acid, mucic acid, 2r,3s,4r,5s-2,3,4,5-tetrahydroxyhexanedioic acid, saccharolactic acid, mucicacid, unii-e149j5otif, d-galactaric acid, galactosaccharic acid, galactarate, meso-galactaric acid
PubChem CID 3037582
ChEBI CHEBI:30852
IUPAC Name (2S,3R,4S,5R)-2,3,4,5-tetrahydroxyhexanedioic acid
SMILES C(C(C(C(=O)O)O)O)(C(C(=O)O)O)O

Specifications

Melting Point ∼210°C (decomposition)
Quantity 100 g
Beilstein 1728117
Merck Index 14,4331
Solubility Information Soluble in water.
Formula Weight 210.14
Percent Purity 98%
Chemical Name or Material Mucic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.