missing translation for 'onlineSavingsMsg'
Learn More

Mucobromic acid, 98+%

Product Code. 11468596
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468596 100 g 100g
11458596 25 g 25g
11478596 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468596 Supplier Thermo Scientific Alfa Aesar Supplier No. A15998.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 488-11-9 | C4HBr2O3- | 256.857 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-11-9
Molecular Formula C4HBr2O3-
Molecular Weight (g/mol) 256.857
MDL Number MFCD00063745
InChI Key NCNYEGJDGNOYJX-IHWYPQMZSA-M
Synonym mucobromic acid, z-2,3-dibromo-4-oxo-2-butenoate, z-2,3-dibromo-4-oxobut-2-enoate, 2z-2,3-dibromo-4-oxobut-2-enoate, z-2,3-bis bromanyl-4-oxidanylidene-but-2-enoate
PubChem CID 6994897
IUPAC Name (Z)-2,3-dibromo-4-oxobut-2-enoate
SMILES C(=O)C(=C(C(=O)[O-])Br)Br

Spécification

Melting Point 121°C to 124°C
Quantity 100 g
UN Number UN3261
Beilstein 1705707
Formula Weight 257.88
Percent Purity ≥98%
Chemical Name or Material Mucobromic acid

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.