missing translation for 'onlineSavingsMsg'
Learn More

Mucobromic acid, 98+%

Product Code. 11478596
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478596 500 g 500g
11458596 25 g 25g
11468596 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478596 Supplier Thermo Scientific Alfa Aesar Supplier No. A15998.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 488-11-9 | C4HBr2O3- | 256.857 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-11-9
Molecular Formula C4HBr2O3-
Molecular Weight (g/mol) 256.857
MDL Number MFCD00063745
InChI Key NCNYEGJDGNOYJX-IHWYPQMZSA-M
Synonym mucobromic acid, z-2,3-dibromo-4-oxo-2-butenoate, z-2,3-dibromo-4-oxobut-2-enoate, 2z-2,3-dibromo-4-oxobut-2-enoate, z-2,3-bis bromanyl-4-oxidanylidene-but-2-enoate
PubChem CID 6994897
IUPAC Name (Z)-2,3-dibromo-4-oxobut-2-enoate
SMILES C(=O)C(=C(C(=O)[O-])Br)Br

Specifications

Melting Point 121°C to 124°C
Quantity 500 g
UN Number UN3261
Beilstein 1705707
Formula Weight 257.88
Percent Purity ≥98%
Chemical Name or Material Mucobromic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.