missing translation for 'onlineSavingsMsg'
Learn More

Mucobromic acid, 99+%

Product Code. 10592521 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10592521 100 g Glass bottle 100g
10618254 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10592521 Supplier Thermo Scientific Acros Supplier No. 128051000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 488-11-9 | C4H2Br2O3 | 257.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-11-9
Molecular Formula C4H2Br2O3
Molecular Weight (g/mol) 257.86
MDL Number MFCD00063745
InChI Key NCNYEGJDGNOYJX-IHWYPQMZSA-M
Synonym mucobromic acid, z-2,3-dibromo-4-oxo-2-butenoate, z-2,3-dibromo-4-oxobut-2-enoate, 2z-2,3-dibromo-4-oxobut-2-enoate, z-2,3-bis bromanyl-4-oxidanylidene-but-2-enoate
PubChem CID 6994897
IUPAC Name (Z)-2,3-dibromo-4-oxobut-2-enoate
SMILES C(=O)C(=C(C(=O)[O-])Br)Br

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 122°C to 125°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula BrC(CHO)=C(Br)CO2H
Quantity 100 g
Beilstein 03, IV, 1721
Formula Weight 257.86
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Mucobromic acid
Show More Show Less
compliance-icons
Product Identifier
  • Mucobromic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.