missing translation for 'onlineSavingsMsg'
Learn More

Mucochloric acid, 99%

Product Code. 10245130 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10245130

Brand: Thermo Scientific Acros 128065000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 87-56-9 | C4H2Cl2O3 | 168.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-56-9
Molecular Formula C4H2Cl2O3
Molecular Weight (g/mol) 168.96
MDL Number MFCD00006966,MFCD00006966
InChI Key LUMLZKVIXLWTCI-IHWYPQMZSA-N
Synonym mucochloric acid, 2,3-dichloromaleic aldehyde acid, kyselina mukochlorova, 2z-2,3-dichloro-4-oxobut-2-enoic acid, 2-butenoic acid, 2,3-dichloro-4-oxo-, 2z, aldehydodichloromaleic acid, dichloromalealdehydic acid, unii-5i5877jhiw, kyselina mukochlorova czech, ccris 6597
PubChem CID 2771871
IUPAC Name (Z)-2,3-dichloro-4-oxobut-2-enoic acid
SMILES OC(=O)C(\Cl)=C(\Cl)C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 125.0°C to 128.0°C
Color White to Beige
Flash Point 186°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula ClC(CHO)=C(Cl)CO2H
Quantity 500 g
Beilstein 03, 727
Merck Index 15, 6383
Solubility Information Solubility in water: soluble in hot water. Other solubilities: slightly soluble in cold water, soluble in hot benzene and alcohol
Formula Weight 168.96
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Mucochloric acid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mucochloric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.