missing translation for 'onlineSavingsMsg'
Learn More

Mucochloric Acid, TRC

Product Code. 30476262
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476262 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30476262 Supplier Toronto Research Chemicals Supplier No. TRCM790800100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mucochloric Acid
CAS 87-56-9
Molecular Formula C4H2Cl2O3
Quantity 100 g
Synonym (Z)-2,3-Dichloro-4-oxo-2-butenoic Acid, Dichloromalealdehydic Acid, 2,3-Dichloromaleic Aldehyde Acid, Dichloromalealdehydic Acid, α,β-Dichloro-β-formylacrylic Acid
InChI Formula InChI=1S/C4H2Cl2O3/c5-2(1-7)3(6)4(8)9/h1H,(H,8,9)/b3-2-
SMILES OC(=O)\C(=C(\Cl)/C=O)\Cl
IUPAC Name (Z)-2,3-dichloro-4-oxobut-2-enoic acid
Molecular Weight (g/mol) 168.96
Formula Weight 167.9381
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.