missing translation for 'onlineSavingsMsg'
Learn More

Melford MUG

Product Code. 30018525
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
25 g
Unit Size:
100mg
1g
250mg
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018525 25 g 25g
30018522 100 mg 100mg
30018523 250 mg 250mg
30018524 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30018525 Supplier Melford Supplier No. M6590025.0
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Fluorescent substrate for β-D-Glucuronidase (GUS) encoded by the gusA gene isolated from E.

Fluorescent substrate for β-D-Glucuronidase (GUS) encoded by the gusA gene isolated from E. coli. Fluorescence assay allows quantitation of GUS activity in protein extracts at peak excitation of 365 nm (UV) and a peak emission of 455 nm (blue).

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.