missing translation for 'onlineSavingsMsg'
Learn More

Manutan Multi-drawer unit, blue, 15 drawers, 940mm x 280mm x 410mm

Product Code. 12875519
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12875519 Supplier Manutan Supplier No. A031825

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Multi-drawer units provide a large storage capacity, using as little floor space as possible. Both unit and drawers are made entirely out of sheet metal. All units are mounted on a black plinth, except for the five drawer unit. The drawer fronts are equipped with chromed label holders and grips.

  • Number of drawers: 15
  • Blue RAL5003
  • Dimensions (H x W x D): 940mm x 280mm x 410mm
  • Drawer dimensions (H x W x D): 51mm x 235mm x 380mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.