Learn More
ratiolab™ Multibox™ Plus Pipette Tip Box
Product Code. 15317417
Change view
Click to view available options
Autoclavable:
Autoclavable
Unit Size:
Each
Product Code. Autoclavable Unit Size
15317417 Autoclavable Each
1 options
Product Code. 15317417 Supplier ratiolab™ Supplier No. 8010161
  • ONLINE EXCLUSIVE PRICE
    11.00 €
    7.16 € / Pack of 1
    Save 3.84 €
    34% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Reusable pipette tip boxes for holding Multitray™ Pipette Tips.
  • For use with the Ratiolab™ MuliTray™ refill systems, Multibox™ plus
  • With dust-proof hinged and sliding lid
  • Completely removable
  • Autoclavable at 121°C, 20 min.
Specifications
Autoclavable Autoclavable
Material Polycarbonate
For Use With (Equipment) Pipette Tips Up to 1200 μL Volume