missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- For safety reasons, MultiDrive TC1 Protective Housing is mandatory to operate MT150 and BT250 tubes (MultiDrive will not start if TC1 isn't attached)
- Solvent resistant
- Dishwasher safe
Specifications
Specifications
| Product Type | Protective Housing for MultiDrive MT150 and BT250 |
| For Use With (Equipment) | IKA MultiDrive Control in combination with MT150 Milling Tube or BT250 Blending Tube |
| Weight (Metric) | 0.365 kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?