missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Multidrop™ 384 Reagent Dispenser

Product Code. 11530725 Shop All Thermo Scientific Products
Change view
Click to view available options
Voltage:
100/120 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
11530725 100/120 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11530725 Supplier Thermo Scientific™ Supplier No. 5840157

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Increase productivity with this reagent dispenser, offering continuous high-speed dispensing of liquids with excellent precision and robot compatibility.

Increase productivity with Thermo Scientific™ Multidrop™ 384 Reagent Dispenser, which offers continuous high-speed dispensing of various liquids with excellent precision and robot compatibility.

Features:

  • Portions a wide range of solutions
  • 96- and 384-well plates across a 5 to 395μL volume range
  • Precise and reproducible dispensing results for pharmaceutical and biotechnology laboratories
  • High-speed dispensing for high assay throughput
  • Easy setup with quick plate and volume selection
  • Easy to integrate with different robotic setups for higher throughput

Convenient, Autoclavable Dispensing Cassette:

  • Eight-channel detachable, autoclavable dispensing cassette to ensure sterile conditions and avoid cross contamination
  • All reagent lines can be backflushed into the reagent bottle, minimizing the loss of expensive reagents

Includes:
One standard tube dispensing cassette (Cat. No. 24072670)

Recommended for:

  • Assay development
  • Primary and secondary screening
  • Compound storage
  • Genomics and proteomics research
  • Cell based assays
  • ELISA assays
TRUSTED_SUSTAINABILITY

Specifications

Accuracy 5µL: ±3% (typical), 20µL: ±2% (typical), 100µL: ±1% (typical)
Product Type Reagent Dispenser
Channels 8
Volume (Metric) 5 to 395 μL
Increments 5 μL
Interface RS-232
Plate Type 96-, 384-well plates
Precision 5μL: CV≤10% (typical), 20μL: CV≤1.5% (typical),
Speed 96-well plate: 20µL in 5 seconds, 50µL in 8 seconds and 384-well plate: 20µL in 20 seconds, 50µL in 25 seconds
Volume (Metric) Dead <7 mL (reagent recovery option)
Width (English) 12.2 in.
Width (Metric) 31 cm
Height (English) 6.1 in.
Height (Metric) 15.5 cm
Depth (English) 12.6 in.
Depth (Metric) 32 cm
Weight (English) 13.7 lb.
Weight (Metric) 6.2 kg
Electrical Requirements 100 to 120V 50/60Hz
Voltage 100/120 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.