missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Multidrop™ Combi Reagent Dispenser

Product Code. 15377400 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Multidrop Combi SMART+
Multidrop Combi w/ SMART 2 Option
Multidrop Combi+
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
15377400 Multidrop Combi w/ SMART 2 Option Each
17801692 Multidrop Combi+ Each
17811692 Multidrop Combi SMART+ Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15377400 Supplier Thermo Scientific™ Supplier No. 5840320

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Meet your lab's specific needs with outstanding flexibility and performance with the easy-to-use Thermo ScientificTM MultidropTM Combi Reagent Dispenser.

Meet your lab's specific needs with outstanding flexibility and performance with the easy-to-use Thermo Scientific™ Multidrop™ Combi Reagent Dispenser, designed to support the varied requirements of pharmaceutical and biotechnology laboratories. It combines the most versatile features for reagent dispensing with excellent performance for drug discovery or genomic and proteomic assays.

The Multidrop Combi Reagent Dispenser handles a wide selection of plates and volume range and provides fast dispensing and high-throughput operation.

  • Provides precise dispensing over a 0.5 to 2500μL range, ensuring reproducible assay data
  • Accommodates microplates from 6 to 1536 wells and plate heights of 5 to 50mm
  • Visual icon-based graphic display makes it easy to use and program, even without training
  • Minimal dead volume and back-flushing reduces reagent costs
  • Easy to use Thermo Scientific FILLit Software provides increased flexibility and functionality
  • Full robotic compatibility gives increased throughput
  • Uses eight-channel detachable and autoclavable dispensing cassettes that are standard across the Multidrop range

Multidrop Combi SMART:

  • Adds an advanced built-in tracing system to the Multidrop Combi
  • Provides improved realiability and cassette lifetime traceability, greatly enhancing user efficiency and reporting capability

Recommended for:

  • Assay development
  • Primary and secondary screening
  • Compound storage
  • Genomics and proteomics research
  • PCR set-up
  • Sequencing set-up
  • Cell based assays
  • Bead based assays
  • ELISA assays
TRUSTED_SUSTAINABILITY

Specifications

Accuracy Small tube dispensing cassette: 2μL, ±10% (typical); 10μL, ±5% (typical); >10μL, ±5% (typical)
Stand tube dispensing cassette: 5μL, ±3%(typical); 20μL, ±2% (typical); 100μL, ±1% (typical); >100μL, ±1% (typical)
Product Type Reagent Dispenser
Description Multidrop Combi w/ SMART 2 Option
Channels 8
Volume (Metric) 0.5 to 2500 μL
Increments Small tube dispensing cassette: 0.5μL increments 0.5-50μL
Stand tube dispensing cassette: 5μL increments 5-2500μL
Interface Serial RS-232, USB
Plate Type 6- to 1536-well plates
Precision Small tube dispensing cassette: 0.5μL, CV ≤10%; 2μL, CV≤5%; 10μL, CV≤3%; >10μL, CV≤3%
Stand tube dispensing cassette: 5μL, CV≤10%; 20μL, CV≤1.5%; 100μL, CV≤1%; >100μL, CV≤1%
Speed 96-well plate: 10μL in 3 sec.; 20μL in 4 sec.; 100μL in 10 sec
384-well plate: 1μL in 5 sec.; 5μL in 5 sec.; 10μL in 6 sec.; 20μL in 9 sec.
1536-well plate: 1μL in 14 sec.; 5μL in 26 sec.
Volume (Metric) Dead <1 mL volume range 0.5to 50 μL; <7 mL volume range >50 μL
Width (English) 14 in.
Width (Metric) 35.5 cm
Height (English) 8.6 in.
Height (Metric) 22 cm
Depth (English) 12.9 in.
Depth (Metric) 33 cm
Weight (English) 20.1 lb.
Weight (Metric) 9.1 kg
Electrical Requirements 100 to 240V 50/60Hz
Voltage 100/240 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.