Learn More
Beschreibung
Accelerate larger scale assays with the superior precision and accuracy of the Thermo Scientific™ Multidrop™ DW Reagent Dispenser, a high-speed reagent dispenser designed for repetitive dispensing of large volumes in pharmaceutical and biotechnology laboratories.
- Precise dispensing ensures high-quality assay results and reproducible data
- Wide volume range provides flexibility and increased speed
- Quick plate and volume selection, combined with easy maintenance, makes assay setup quick and easy
- Dispenser uses an eight-channel detachable and autoclavable dispensing cassette to ensure sterile conditions and to avoid cross contamination
- All reagent lines can be backflushed into the reagent bottle, minimizing the loss of expensive reagents
Includes:
One standard tube dispensing cassette (Cat. No. 24072670).
Recommended for:
- Cell based assays
- Compound storage
- Genomics and proteomics research
- ELISA assays
- Microbiological tests
Spezifikation
Spezifikation
| Accuracy | 20µLl: ±2% (typical); 100µL: ±1% (typical); 900µL: ±1% (typical) |
| Product Type | Reagent Dispenser |
| Channels | 8 |
| Volume (Metric) | 20 to 995 μL |
| Increments | 5 μL |
| Interface | RS-232 |
| Plate Type | 96-well plates; 96-deep well plates; 1.1mL tubes in 96 tube rack; Micronic 96-place tube racks |
| Precision | 20μL: CV≤1.5% (typical), 100μL: CV≤1% (typical), 900μL: CV≤0.5% (typical) |
| Speed | 96-well plate: 20µL in 5 seconds; 50µL in 8 seconds; 300µL in 26 seconds; 900 µL in 74 seconds |
| Volume (Metric) Dead | <7 mL |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.