missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Multidrop™ Pico Digital Dispenser

Product Code. 31337546 Shop All Thermo Scientific Products
Change view
Click to view available options
Model:
Multidrop Pico 1 Digital Dispenser
Multidrop Pico 8 Surfactant Free Digital Dispenser
Dimensions (L x W x H):
38.5 x 28 x 19 cm (15 in x 11 in x 7.5 in)
47 x 38 x 23 cm (18.5 x 15 x 9.1 in)
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Dimensions (L x W x H) Unit Size
31337546 Multidrop Pico 8 Surfactant Free Digital Dispenser 47 x 38 x 23 cm (18.5 x 15 x 9.1 in) -
31722884 Multidrop Pico 8 Surfactant Free Digital Dispenser 47 x 38 x 23 cm (18.5 x 15 x 9.1 in) Each
16671296 Multidrop Pico 1 Digital Dispenser 38.5 x 28 x 19 cm (15 in x 11 in x 7.5 in) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31337546 Supplier Thermo Scientific™ Supplier No. 5840700
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Multidrop™ Pico Digital Dispensers provide high-precision, non-contact dispensing from 8 pL to 10 μL, using up to ten times less reagent than manual pipetting. Ideal for miniaturizing assays, they ensure accurate results with user-friendly software.

The Multidrop™ Pico Digital Dispensers offer enhanced dispensing precision with volumes as low as 8 pL, using up to ten times less reagent than manual pipetting, reducing costs and waste. Designed for assay miniaturization, they boost productivity and improve accuracy, ideal for qPCR, dose response curves, drug screening assays, and enzyme profiling. The intuitive Multidrop™ PicoIT software supports complex experimental setups, enhancing productivity and data integrity while conserving resources and improving operational efficiency.

Features:

  • Advanced Assay Miniaturization: Digital dispensing technology enables direct dilution and low dead volume with precise fluid delivery as low as 11 pL enabling up to 10-fold reduction in reagent and sample consumption.
  • Enhanced Workflow Efficiency and Sample Integrity: Non-contact disposable dispensing cartridges eliminate cross-contamination and carry-over. With a 40x increase in processing speed, typical dose experiments are completed in just 15 seconds, compared to 10 minutes with analog serial dilution. This minimizes manual pipetting steps and reduces the risk of repetitive strain injuries.
  • Intuitive Software: Multidrop™ PicoIT software provides flexible and intuitive experiment design capabilities for complex plate layouts. It supports sophisticated assay configurations while maintaining user-friendly operation through simplified setup procedures. Application-focused wizards and pre-programmed protocol templates for qPCR master mixes provide automated fluid customization for each qPCR component. Complete traceability functionality tracks reagent amounts and dispensing times for each well after every run.
  • Wide Range of Applications: The Multidrop™ Pico digital dispensers support a variety of fluid types, including master mixes, DMSO-based and aqueous fluids, with or without small molecule compounds or biomolecules. The Multidrop™ Pico 8 Surfactant Free (Pico 8 sf) can dispense aqueous fluids without surfactant using surfactant free cartridges. The digital dispensers automate complex protocols such as drug synergy/combination studies and multi-component titrations. They are ideal for applications including qPCR, drug combination studies, DMPK, assay development, biochemical assays, secondary screening, cell-based assays, spotting, enzyme profiling, synthetic biology, antibody therapies, and SAR.
  • Instrument Options and Capabilities: Choose between two instrument options: the Pico 1, which uses 1-channel cartridges, and the Pico 8 sf, which uses 4- or 8-channel cartridges. The Pico 8 sf is specifically designed for complex multi-reagent/sample assays in high-density formats up to 1536-well plates.

Service Plan

Yes

TRUSTED_SUSTAINABILITY

Specifications

Product Type Digital Dispenser
Channels 4,8
Volume (Metric) 8 pL to 10 μL
For Use With (Application) Drug combination studies, Biochemical assays, Cell-based assays, DMPK, Secondary screening, Synthetic chemistry, Assay development, qPCR, Enzyme profiling, Antibody therapies, SAR, and Spotting
For Use With (Equipment) 4-channel, 200 μL Standard Cartridges; 8-channel, 20 μL Standard Cartridges; 4-channel, 200 μL Surfactant Free Cartridges; 8-channel ,20 μL Surfactant Free Cartridges
Increments As low as 8 pL
Interface USB, PC Control
Model Multidrop Pico 8 Surfactant Free Digital Dispenser
Plate Type 12, 24, 48, 96, 384, 1536 well plate, Plate height: 6-47 mm
Precision ≤ 8%
Software Multidrop PicoIT Software
Dimensions (L x W x H) 47 x 38 x 23 cm (18.5 x 15 x 9.1 in)
Weight (English) 31.1 lb
Weight (Metric) 14.1 kg
Electrical Requirements 100 to 240 V 50/60Hz
Voltage 100/240 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.