missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek MultiFlo FX Multi-Mode Dispenser
GREENER_CHOICE

Product Code. 17217829 Shop All Agilent BioTek Products
Change view
Click to view available options
Model:
MFXP1
MFXP2
MFXP2R
Includes:
1 module arm, 1 peristaltic dispense pump and 1 peri-pump cassette with plastic tips
2 module arms, 1 peristaltic dispense pump and 1 peri-pump cassette with plastic tips
RAD single channel dispenser, 2 module arms, 2 peristaltic dispense pumps, 1 RAD manifold and tubing
Unit Size:
Each
Product Code. Model Includes unitSize
17217829 MFXP2 2 module arms, 1 peristaltic dispense pump and 1 peri-pump cassette with plastic tips Each
17207829 MFXP1 1 module arm, 1 peristaltic dispense pump and 1 peri-pump cassette with plastic tips Each
17227829 MFXP2R RAD single channel dispenser, 2 module arms, 2 peristaltic dispense pumps, 1 RAD manifold and tubing Each
3 options
This item is not returnable. View return policy

Product Code. 17217829

Brand: Agilent BioTek MFXP2SN

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MultiFlo FX is an automated multi-mode reagent dispenser for 6- to 1536-well microplates.

MultiFlo FX is an automated multi-mode reagent dispenser for 6- to 1536-well microplates. The system incorporates several unique technologies in its modular design, such as Parallel Dispense, random access dispense (RAD) and patented Automated Media Exchange (AMX) modules

The technologies in MultiFlo FX microplate dispenser enable many liquid handling applications from 2D and 3D cell culture to concentration normalization assays, ELISA, bead-based assays and more. A fully configured MultiFlo FX replaces up to five liquid handlers, saving space, time, and instrumentation budgets. MultiFlo FX delivers complete workflow automation in cell imaging and biochemical applications.

TRUSTED_SUSTAINABILITY

Specifications

Accuracy ±5% at 1 μL, ±2.0% at 5 μL, ±2.0% at 10 μL
Product Type Microplate Dispenser
Certifications/Compliance CE and TUV marked. RoHS compliant.
Volume (Metric) 500 nL to 3000 μL/well, 1 μL increments
For Use With (Application) Primary/secondary screening assays, compound storage, genomics and proteomics research, ELISA, cell based washing, fixing, staining, volume or concentration normalization
For Use With (Equipment) BioStack and third party automation compatible; BioSpa 8 Automated Incubator compatible
Includes 2 module arms, 1 peristaltic dispense pump and 1 peri-pump cassette with plastic tips
Interface Color touch screen; Create, edit or run multiple protocols onboard
Model MFXP2
Plate Type Dispensing (peri pump and syringe): 96-, 384- and 1536-well standard, deep and PCR plates; 6-. 12-, 24- and 48-well plates (dispense tip configurable); Washing: 96-. 384-well standard plates; 6-, 12-, 24-,and 48-well plates (with compatible manifold configuration)
Precision <5% CV at 1 μL, <2.5% CV at 5 μL, <2.0% CV at 10 μL
Software LHC2 Software; LHC2 Secure for 21 CFR Part 11 compliance (option); SiLA Compliant driver (option)
Speed 96 wells, 5 μL cass, 10 μL/ well: 3 s, 96 wells, 5 μL cass, 20 μL/ well: 3.5 s, 384 wells, 5 μL cass, 5 μL/ well: 6.5 s, 384 wells, 10 μL cass, 10 μL/ well: 8 s, 384 wells, 1 μL cass, 1 μL/ well: 6 s, 1536 wells, 1 μL cass, 1 μL/ well: 21 s
Width (English) 17.2 in.
Width (Metric) 43.5 cm
Height (English) 8 in.
Height (Metric) 20.3 cm
Depth (English) 11.8 in.
Depth (Metric) 29.2 cm
Weight (English) 14.3 lb.
Weight (Metric) 6.5 kg
Electrical Requirements 90 W max.
Voltage 100/240 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.