Learn More
WTW™ MultiLab™ Pro 20 IDS S 2
Product Code. 30929046
Change view
Click to view available options
Channels:
2
Unit Size:
Each
Product Code. Channels unitSize
30929046 2 Each
1 options
Product Code. 30929046 Supplier WTW™ Supplier No. 1FD76C
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
Xylem’s MultiLab™ Pro IDS is a modern high-performance multiparameter device with 2 galvanically isolated measuring channels for pH, ORP, mV, conductivity, oxygen, ISE and turbidity measurement. Up to 2 identical or different parameters can be measured simultaneously.
The MultiLab™ Pro IDS from Xylem is a modern multi-parameter measuring device with up to two measurement channels.
  • Ease of use
  • LIMS ready
  • GxP compliance
  • Compact and functional design
Highlights:
  • Measures pH, ORP, conductivity, dissolved oxygen, ISE (coming soon) and turbidity
  • Wireless-ready for IDS modules
  • Digital sensor recognition
  • Simultaneous measurement with up to two independent measuring channels
  • GxP-compliant
  • Brilliant touch display
  • Preset and definable buffer sets
  • QSC calibration
  • AutoRead function
  • LIMS ready
  • Export to PDF/A, JSON and CSV
  • Special touchscreen glass can be operated with gloves and resistant to alcohol and disinfectants
Scope of delivery:
  • Device with stand and power supply, SenTix™ 980 (pH), TetraCon™ 925 (COND), accessories
TRUSTED_SUSTAINABILITY
Specifications
Calibration pH
Certifications/Compliance CE Certified
Channels 2
Display Type Color TFT
IP Rating IP42
Measurement Modes pH, Redox, mV, Conductivity, Oxygen
Range (Conductivity) 0.001 μS/cm to 200 μS/cm
Range (pH) 0.000 to 14.000
Resolution (ISE) depending on Sensor
Resolution (pH) depending on Sensor
Temperature Range (Metric) depending on Sensor
Log Function Type Manual or Interval
Product Type Multiparameter Laboratory Measuring Device
Voltage 115/230 VAC