missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Multiparameter Chemical Test Kit
Description
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
- Complete setup: All required materials are included, such as beakers, syringes, a case and pre-measured reagent bottles and packets
- High resolution readings for fast, accurate determinations
- Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
Specifications
| For Use With (Application) | Environmental Monitoring |
| For Use With (Equipment) | Water samples |
| Includes | All necessary reagents and accessories for each test, pHep™ electronic pH tester, hard carrying case, and instructions |
| No. of Tests | 6 parameters x 100 tests each |
| Quantity | 1 Kit |
| Range | Various |
| Sufficient For | Testing of: pH, acidity, alkalinity, carbon dioxide, hardness, dissolved oxygen |
| Test Method | Titration, colorimetry, and direct measurement |
| Type | Environmental Monitoring Test Kit |
Safety and Handling
Product Identifier
- 1 SET ECOLOGY TEST KIT HI4814
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 A
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H226-Flammable liquid and vapour.
- H290-May be corrosive to metals.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H350-May cause cancer.
- H361f-Suspected of damaging fertility.
- H372-Causes damage to organs.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- EUH031-Contact with acids liberates toxic gas.
- EUH032-Contact with acids liberates very toxic gas.
- EUH210-Safety data sheet available on request.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use powder to extinguish.
- P390-Absorb spillage to prevent material damage.
- P391-Collect spillage.
- MIXTURE LIST-Contain: Sodium hydroxide, Potassium iodide, Sodium azide, Propionicacid, Hydrochloric acid, Manganese(II) s
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?