Learn More
VCAM-1/CD106 Rat anti-Mouse, Alexa Fluor 647, Clone: 112734, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems FAB6432R-100UG
Encompass
Product Code. 15738153
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Mouse samples. It is validated for Flow Cytometry.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)