missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Edmont™ Marigold™ Multiplus 40 Gloves
Click to view available options
Size:
8
9
10
11
Unit Size:
12 pairs
Description
- Fully coated in high performance PVC/nitrile
- 100% cotton interlock liner on palm, thumb and forefinger
- Resistance to abrasion and tearing
- Superior plasticized plastoprene integral base coating, made of mix of resins and plasticizers with incorporated elastomers
- Anatomical shape for increased dexterity and comfort
- Ieal for tasks involving mechanical and chemical hazards: maintenance, cleaning, petrochemical plants, factories.
Specifications
Specifications
| Description | Not Anti-static, Not silicone-free |
| Color | Blue |
| Latex Free | No |
| Material | PVC |
| Size | 8 |
| Glove Cuff Style | Gauntlet |
| Liner Material | Interlock cotton |
| Resistant To | Chemical and liquid protection |
| Series | Multiplus 40 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction