Learn More
Brady™ Multipurpose Vinyl Label Tape for M6/M7 Printers
With availability in 12 colors, colored vinyl labels are ideal for product, component, rating plate and general identification where various color options is required.
Brand:  Brady™ 174309
 View more versions of this product
Product Code. 17838744
  • ONLINE EXCLUSIVE PRICE
    85.70 €
    82.73 € / Pack of 1
    Save 2.97 €
    3% Off
Estimated Shipment: 11-12-2026
Add to basket
Description
Description
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty, and harsh environment surfaces, and can be applied in cold temperatures
  • Designed for use in ambient conditions with limited solvent exposure
  • RoHS compliant to 2011/65/EU (Pass 2015/863)
  • Made of B-439 vinyl material
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
Black
Continuous
Single-side Printable
Label
Barcode Labeling, Equipment Labeling, Electrical Labeling, General Identification, Inventory and Inspection Labeling
No
Rectangle
-70°C to +80°C
0.107 mm
50 ft.
1 in.
M6C-1000-439-BK
ASTM D 1000, ASTM D 2979, ASTM G 155
None
Gloss
Vinyl
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
50 ft.
-94°F to +176°F
0.0042 in.
50 ft. x 1 in.
15.24 m
25.4 mm