missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ MultiRack Empty Rack

Product Code. 16382237 Shop All Axygen Scientific Products
Change view
Click to view available options
For Use With (Equipment):
10 μL XT, 200 μL, 200 μL NX, 300 μL
200 μL XT, 200 μL XT, 300 μL XT, 1000 μL XT
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16382237 200 μL XT, 200 μL XT, 300 μL XT, 1000 μL XT Pack of 5
16372237 10 μL XT, 200 μL, 200 μL NX, 300 μL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16382237 Supplier Axygen™ Supplier No. MRRB
  • ONLINE EXCLUSIVE PRICE
    114.00 €
    111.64 € / Pack of 5
    Save 2.36 €
    2% Off
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in Bulk, Racked, Filter and Reload packaging styles to meet the variety of needs in the lab setting.

  • Detectable DNase/RNase/Human DNA/PCR inhibitor free
  • Nonpyrogenic
  • Manufactured in an ISO certified facility in the USA
  • Fully autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 13845
Autoclavable Autoclavable
Color Clear
Material Polyethylene
Sterility Non-sterile
For Use With (Equipment) 200 μL XT, 200 μL XT, 300 μL XT, 1000 μL XT
No. per Case 1/Cs.
No. per Pack 5/Pk.
Quantity 5/pk, 1 pk/cs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.