missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Multiskan™ FC Microplate Photometer Lamp

Product Code. 11861575 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
Lamp 6V/10W
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
11861575 Lamp 6V/10W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11861575 Supplier Thermo Scientific™ Supplier No. 1410101
Discount Lab Supplies
Use promo code 28170
View all promotions
Estimated Shipment: 11-12-2026
Add to basket
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Règlementaire : Cet article est soumis à éco-contribution.

Measure absorbance in a wide variety of research and clinical applications with the easy-to-use Thermo Scientific™ Multiskan™ FC Microplate Photometer.

Measure absorbance in a variety of research and clinical applications with the easy-to-use Thermo Scientific™ Multiskan™ FC Microplate Photometer. The Multiskan FC provides proven performance and reliable results through built-in self diagnostics, IQ/OQ/PQ and verification tools. The visual user interface and multiple language capability ensures excellent usability. The Multiskan FC (IVD model) with internal software conforms to the European IVD (In Vitro Medical Device) directive, making it ideal for qualitative and quantitative ELISA applications in clinical laboratories. Simplify your ELISA workflow by using our eGuide for all your ELISA needs.

Features:
  • Wide wavelength range of 340 to 850nm
  • Fast reading of both 96- and 384-well plates
  • Shaking and incubation up to 50°C for temperature critical assays
  • Ease of use through the large color screen and a variety of language versions
  • Superior usability and logical workflow with Thermo Scientific™ SkanIt™ Software
  • USB port for easy data transfer

Recommended for:

Immunoassays (ELISA), protein assays, endotoxins, cytotoxicity and proliferation assays, enzyme assays, growth curves

TRUSTED_SUSTAINABILITY

Spécification

Product Type Lamp Assembly
For Use With (Equipment) Multiskan FC
Product Line Multiskan
Includes Lamp 6V/10W
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.