missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ multiSUB™ Midi Horizontal Gel System

Product Code. 12397277 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Sample Size:
50
100
Gel Dimensions:
W x L: 10 x 10 cm
W x L: 10 x 7 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sample Size Gel Dimensions Unit Size
12397277 100 W x L: 10 x 10 cm Each
12307287 50 W x L: 10 x 7 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12397277
  • ONLINE EXCLUSIVE PRICE
    542.00 €
    371.51 € / Pack of 1
    Save 170.49 €
    31% Off
View all promotions
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Designed for routine horizontal gel electrophoresis.

  • Suitable for rapid electrophoresis
  • Low buffer volumes
  • Runs about 50 to 100 samples

Specifications

Type Horizontal Gel Electrophoresis System
Gel Dimensions W x L: 10 x 10 cm
Includes multiSUB™ Midi electrophoresis unit, 10 x 10cm UV Tray, 2 x 16 sample combs, loading guides and dams
Sample Size 100
Volume (English) Buffer 0.07 gal.
Volume (Metric) Buffer 300 mL
Length (English) 4.92 in.
Length (Metric) 12.5 cm
Width (English) 8.66 in.
Width (Metric) 22 cm
Height (English) 3.54 in.
Height (Metric) 9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.