missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals MULTISURE™ HIV 1/2 Confirmatory Test Controls

Product Code. 31605224 Shop All MP Biomedicals Products
Change view
Click to view available options
Detectable Analytes:
HIV-1/2
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes Unit Size
31605224 HIV-1/2 Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31605224 Supplier MP Biomedicals Supplier No. 90030006
 Restricted Item
Estimated Shipment: 15-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to monitor and validate the performance of the MULTISURE™ HIV 1/2 confirmatory test, helping to ensure accurate and reliable results

  • MULTISURE™ HIV 1/2 Confirmatory Test Controls quality control materials designed to be used with the MULTISURE™ HIV 1/2 Confirmatory Test system by MP Biomedicals / MP Diagnostics.
  • They are typically used by laboratory professionals as part of quality control procedures for example when opening a new lot of test kits, upon receipt of a new shipment, or at regular intervals as specified by local laboratory requirements.
  • Typically produces confirmatory results within approximately 30 minutes and can be read manually or with compatible readers.

Specifications

CE Marker CE-IVD
Detectable Analytes HIV-1/2
Type Test Controls
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.