missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals MULTISURE HIV Rapid Test (CE)

Product Code. 31605225 Shop All MP Biomedicals Products
Change view
Click to view available options
Detectable Analytes:
HIV-1 gp120, HIV-1 gp41, HIV-1 p24, HIV-2 gp36
Unit Size:
20 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes Unit Size
31605225 HIV-1 gp120, HIV-1 gp41, HIV-1 p24, HIV-2 gp36 20 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31605225 Supplier MP Biomedicals Supplier No. 43030020
 Restricted Item
Estimated Shipment: 15-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A rapid diagnostic test, developed to detect antibodies specific to HIV-1 gp120, HIV-1 gp41, HIV-1 p24 (also react with HIV-2) and HIV-2 gp36 antigens

  • The MP Diagnostics MULTISURE™ HIV Rapid Test is a rapid diagnostic assay.
  • It is designed to detect antibodies specific to the following HIV antigens:
    • HIV-1 gp120
    • HIV-1 gp41
    • HIV-1 p24 (also reactive with HIV-2)
    • HIV-2 gp36
  • The test can be used with human serum, plasma, finger-pricked whole blood, or whole blood containing anticoagulants.
  • Usage Statement: For sale outside the US only.

Specifications

CE Marker CE-IVD
Detectable Analytes HIV-1 gp120, HIV-1 gp41, HIV-1 p24, HIV-2 gp36
Type HIV Rapid Test
compliance-icons
Product Identifier
  • Rapid Test Kits
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.