missing translation for 'onlineSavingsMsg'
Learn More

Murexide, For Complexometry, Honeywell Fluka™

Product Code. 15673950 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
10 g
25 g
Packaging:
Glass Bottle
Unit Size:
10g
25g
5g
This item is not returnable. View return policy

Product Code. 15673950

Brand: Honeywell-Fluka 3341410G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

for complexometry

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3051-09-0
Molecular Formula C8H3N5O6-2
Molecular Weight (g/mol) 265.141
MDL Number MFCD00012777
InChI Key MUMMBXDKKJEASE-UHFFFAOYSA-L
PubChem CID 51381221
IUPAC Name 6-oxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1H-pyrimidine-2,4-diolate
SMILES C1(=C(N=C(NC1=O)[O-])[O-])N=C2C(=O)NC(=O)NC2=O

Specifications

Melting Point >300°C
UN Number NONH for all modes of transport
Beilstein 3831445
Formula Weight 284.19g/mol
Packaging Glass Bottle
Color Red
Quantity 10 g
Physical Form Powder
Chemical Name or Material Murexide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.