missing translation for 'onlineSavingsMsg'
Learn More

Murexide, For Complexometry, Solstice

Product Code. 15683950 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Packaging:
Glass Bottle
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15683950 25 g Glass Bottle 25g
15693950 5 g Glass Bottle 5g
15673950 10 g Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15683950 Supplier Honeywell-Fluka Supplier No. 3341425G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Currently Unavailable: We have no information when this product will be available.
This item is not returnable. View return policy

for complexometry

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3051-09-0
Molecular Formula C8H3N5O6-2
Molecular Weight (g/mol) 265.141
MDL Number MFCD00012777
InChI Key MUMMBXDKKJEASE-UHFFFAOYSA-L
PubChem CID 51381221
IUPAC Name 6-oxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1H-pyrimidine-2,4-diolate
SMILES C1(=C(N=C(NC1=O)[O-])[O-])N=C2C(=O)NC(=O)NC2=O

Specifications

Melting Point >300°C
UN Number NONH for all modes of transport
Beilstein 3831445
Formula Weight 284.19g/mol
Packaging Glass Bottle
Color Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Murexide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.