missing translation for 'onlineSavingsMsg'
Learn More

Murexide

Product Code. 11408177
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11408177 50 g 50g
11498167 10 g 10g
11418177 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408177 Supplier Thermo Scientific Alfa Aesar Supplier No. A17540.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3051-09-0 | C8H3N5O6-2 | 265.141 g/mol

Murexide is used as a complexometric indicator in analytical chemistry for complexometric titrations of calcium, copper nickel and cobalt. It is an inhibitor of dihydrofolate reductase. Further, it acts as an enhancer of sonochemical destruction of chlorinated hydrocarbon pollutants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Murexide is used as a complexometric indicator in analytical chemistry for complexometric titrations of calcium, copper nickel and cobalt. It is an inhibitor of dihydrofolate reductase. Further, it acts as an enhancer of sonochemical destruction of chlorinated hydrocarbon pollutants.

Solubility
Soluble in water and 1M sodium hydroxide solution. Insoluble in alcohol.

Notes
Incompatible with strong oxidizing agents.                  
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3051-09-0
Molecular Formula C8H3N5O6-2
Molecular Weight (g/mol) 265.141
MDL Number MFCD00012777
InChI Key MUMMBXDKKJEASE-UHFFFAOYSA-L
Synonym murexide, ammonium purpurate acid, 6-oxo-5-2,4,6-trioxo-1,3-diazinan-5-ylidene amino-1h-pyrimidine-2,4-diolate, 5-hexahydro-2,4,6-trioxo-5-pyrimidinyl imino-2,4,6 1h,3h,5h-pyrimidinetrione ammonium salt, 6-oxidanylidene-5-2,4,6-tris oxidanylidene-1,3-diazinan-5-ylidene amino-1h-pyrimidine-2,4-diolate
PubChem CID 51381221
IUPAC Name 6-oxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1H-pyrimidine-2,4-diolate
SMILES C1(=C(N=C(NC1=O)[O-])[O-])N=C2C(=O)NC(=O)NC2=O

Specifications

Melting Point >300°C
Quantity 50 g
Beilstein 3582175
Merck Index 14,6306
Solubility Information Soluble in water and 1M sodium hydroxide solution. Insoluble in alcohol.
Formula Weight 284.19
Chemical Name or Material Murexide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.