missing translation for 'onlineSavingsMsg'
Learn More

Murexide

Product Code. 11498167
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498167 10 g 10g
11408177 50 g 50g
11418177 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498167 Supplier Thermo Scientific Alfa Aesar Supplier No. A17540.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 3051-09-0 | C8H3N5O6-2 | 265.141 g/mol

Murexide is used as a complexometric indicator in analytical chemistry for complexometric titrations of calcium, copper nickel and cobalt. It is an inhibitor of dihydrofolate reductase. Further, it acts as an enhancer of sonochemical destruction of chlorinated hydrocarbon pollutants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Murexide is used as a complexometric indicator in analytical chemistry for complexometric titrations of calcium, copper nickel and cobalt. It is an inhibitor of dihydrofolate reductase. Further, it acts as an enhancer of sonochemical destruction of chlorinated hydrocarbon pollutants.

Solubility
Soluble in water and 1M sodium hydroxide solution. Insoluble in alcohol.

Notes
Incompatible with strong oxidizing agents.                  
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3051-09-0
Molecular Formula C8H3N5O6-2
Molecular Weight (g/mol) 265.141
MDL Number MFCD00012777
InChI Key MUMMBXDKKJEASE-UHFFFAOYSA-L
Synonym murexide, ammonium purpurate acid, 6-oxo-5-2,4,6-trioxo-1,3-diazinan-5-ylidene amino-1h-pyrimidine-2,4-diolate, 5-hexahydro-2,4,6-trioxo-5-pyrimidinyl imino-2,4,6 1h,3h,5h-pyrimidinetrione ammonium salt, 6-oxidanylidene-5-2,4,6-tris oxidanylidene-1,3-diazinan-5-ylidene amino-1h-pyrimidine-2,4-diolate
PubChem CID 51381221
IUPAC Name 6-oxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1H-pyrimidine-2,4-diolate
SMILES C1(=C(N=C(NC1=O)[O-])[O-])N=C2C(=O)NC(=O)NC2=O

Specifications

Melting Point >300°C
Beilstein 3582175
Merck Index 14,6306
Solubility Information Soluble in water and 1M sodium hydroxide solution. Insoluble in alcohol.
Formula Weight 284.19
Quantity 10 g
Chemical Name or Material Murexide

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.