missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ (+)-Muscarine iodide

Product Code. 18722664
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18722664 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18722664 Supplier R&D Systems™ Supplier No. 3074/10
  • ONLINE EXCLUSIVE PRICE
    386.73 €
    346.50 € / 10mg
    Save 40.23 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Muscarinic receptor agonist

Muscarinic acetylcholine receptor agonist; mimics the activity of acetylcholine.
TRUSTED_SUSTAINABILITY

Specifications

CAS 24570-49-8
Quantity 10 mg
Product Line Tocris
Molecular Formula C9H20INO2
Formula Weight 301.17
Purity >98%
Molecular Weight (g/mol) 301.168
Chemical Name or Material (+)-Muscarine iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.