missing translation for 'onlineSavingsMsg'
Learn More

(+)-Muscarine Iodide, TRC

Product Code. 30012027
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012027 50 mg 50mg
30007021 5 mg 5mg
30008392 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012027 Supplier Toronto Research Chemicals Supplier No. TRCM81400050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24570-49-8
Molecular Formula C9 H20 N O2 . I
Molecular Weight (g/mol) 301.17
IUPAC Name [(2S,4R,5S)-4-hydroxy-5-methyloxolan-2-yl]methyl-trimethylazanium;iodide
SMILES [I-].C[C@@H]1O[C@H](C[N+](C)(C)C)C[C@H]1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H20NO2.HI/c1-7-9(11)5-8(12-7)6-10(2,3)4;/h7-9,11H,5-6H2,1-4H3;1H/q+1;/p-1/t7-,8-,9+;/m0./s1
Formula Weight 301.0539
Chemical Name or Material (+)-Muscarine Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.