missing translation for 'onlineSavingsMsg'
Learn More

Myclobutanil, TRC

Product Code. 30132887
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132887 1 g 1g
17902235 100 mg 100mg
17933185 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132887 Supplier Toronto Research Chemicals Supplier No. TRCM8314001G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88671-89-0
Molecular Formula C15 H17 Cl N4
Molecular Weight (g/mol) 288.78
Synonym 1H-1,2,4-Triazole-1-propanenitrile, α-butyl-α-(4-chlorophenyl)-, α-Butyl-α-(4-chlorophenyl)-1H-1,2,4-triazole-1-propanenitrile, (±)-Myclobutanil, Deccotanil, Eagle, Fungiben, HOE 39304F, Laredo 25EC, Laredo 25EW, Myclobutanil, Mycloss Xtra, Nova, Nova (pesticide), Nova 40W, Nova W, Nu-Flow M, RH 3866, Rally, Spectracide Immunox Multi-Purpose Fungicide, Systhane, Systhane 12E, Systhane 40W, Systhane 6 Flo
IUPAC Name 2-(4-chlorophenyl)-2-(1,2,4-triazol-1-ylmethyl)hexanenitrile
SMILES CCCCC(Cn1cncn1)(C#N)c2ccc(Cl)cc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H17ClN4/c1-2-3-8-15(9-17,10-20-12-18-11-19-20)13-4-6-14(16)7-5-13/h4-7,11-12H,2-3,8,10H2,1H3
Formula Weight 288.1142
Percent Purity >95%
Chemical Name or Material Myclobutanil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.