missing translation for 'onlineSavingsMsg'
Learn More

MP BIOMEDICALS Myco-Out Mycoplasma Removal Spray

Product Code. 30130211 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 x 450 mL
4 x 450 mL
Unit Size:
1.8 L
450mL
This item is not returnable. View return policy

Product Code. 30130211

Brand: MP BIOMEDICALS 093050853X4

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Myco-Out Mycoplasma Removal Spray is easy to use, fast and non-toxic, highly effective, and features a broad antimicrobial spectrum that eliminates Mycoplasma and inactivates various microorganisms.

Mycoplasma is a frequent contaminant of cell cultures. It is the smallest and the simplest prokaryotic organism without cell walls. Therefore, it can pass through 0.22 μm filters easily. Moreover, the common antibiotics which target the cell wall, such as penicillin or β-lactam antibiotics, are ineffective against mycoplasmas. Mycoplasma contamination can severely affect the state of cells, causing changes in gene expression and metabolic characteristics, decelerated cell growth, abnormal cell differentiation, and cell death. It also severely affects cellular functions.

MP Biomedicals Myco-Out Mycoplasma Removal Spray is a ready-to-use peptide surfactant, which can quickly, efficiently and non-toxically eliminate mycoplasma from working surroundings such as:

  • Benches
  • Biosafety cabinets
  • Laminar flow cabinets
  • Cell incubators
  • Pipettes
  • Liquid nitrogen tanks
  • Door handles, etc.

It also has a broad-spectrum antimicrobial disinfection effect.

Features:

  • Easy to Use: Myco-Out Mycoplasma Removal Spray simplifies Mycoplasma removal with a user-friendly, ready-to-use spray, eliminating the need for tedious methods like fumigation and alcohol wiping
  • Fast and Non-Toxic: The mycoplasma-specific peptide surfactant derived from Bacillus subtilis works in just 5 to 10 minutes without harming cells
  • Highly Effective: Efficacy studies confirm its effectiveness in removing Mycoplasma from surfaces like laminar flow cabinets coated with Mycoplasma-positive HeLa cell culture
  • Broad Antimicrobial Spectrum: It eliminates Mycoplasma and inactivates various microorganisms, including bacteria, viruses, and fungal spores
  • Safe to Use: The product is safe for users and the environment

Application Notes:

Myco-Out Mycoplasma Removal Spray is a ready-to-use peptide surfactant intended to:

  • Quickly
  • Efficiently
  • and non-toxically eliminate mycoplasma from working surroundings

Usage Statement:

Research Use Only (RUO)

TRUSTED_SUSTAINABILITY

Specifications

Antibiotic Spectrum Broad spectrum
Classification Chemically-defined
Form Liquid
Product Type Mycoplasma Removal
Reagent Type Surfactant
Sterility Non-sterile
Quantity 4 x 450 mL
compliance-icons compliance-icons
Product Identifier
  • Myco-Out Mycoplasma Removal Spray
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Isopropanol, ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.