missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Myco-Visible Mycoplasma ​PCR Detection

Product Code. 31759330 Shop All MP Biomedicals Products
Change view
Click to view available options
No. of Tests:
100 Tests
20 Tests
Unit Size:
100 tests
20 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests unitSize
31759330 20 Tests 20 tests
31759329 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31759330 Supplier MP Biomedicals Supplier No. 093051401

Please to purchase this item. Need a web account? Register with us today!

Standardized and rapid detection, completes assay workflow in under 3 hours for efficient mycoplasma screening

  • The Myco-Visible Mycoplasma PCR Detection Kit provides a highly sensitive and specific method for detecting mycoplasma contamination in cell cultures.
  • Capable of identifying a wide range of mycoplasma species such as
    • Mycoplasma orale
    • Mycoplasma hyorhinis.
  • The kit delivers results in as little as 3 hours. With a detection limit of 10 CFU/mL, it ensures reliable identification of contamination at the genetic level.
  • The kit includes:
    • an internal control to differentiate valid results from PCR inhibition
    • a positive control for verification.
  • Compliant with EP 2.6.7, and JP 17, this PCR-based solution offers faster and more efficient
  • Alternative to traditional culture methods for mycoplasma testing.
TRUSTED_SUSTAINABILITY

Specifications

PCR Method Standard PCR
Reaction Speed 120 min.
No. of Tests 20 Tests
Product Type Mycoplasma Detection
For Use With (Application) Mycoplasma Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.