missing translation for 'onlineSavingsMsg'
Learn More

MP BIOMEDICALS Myco-Visible Mycoplasma Rapid Test Kit

Product Code. 30928581 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
24 Tests
5 Tests
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30928581 5 Tests 1 set
30928578 24 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. Rückgaberichtlinie anzeigen
Artikelnummer. 30928581 Lieferant MP BIOMEDICALS Lieferanten-Nr. 09305090S
  • ONLINE EXCLUSIVE PRICE
    113.00 €
    107.10 € / 1 sets
    Save 5.90 €
    5% Off
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Edge
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rapid, easy mycoplasma detection for routine cell culture monitoring; broad-species coverage and simple, equipment-free workflow.

  • Detects mycoplasma contamination with a sensitivity as low as 10 CFU/mL or 10 fg of genomic DNA
  • Effective against a broad range of mycoplasma species, including common ones like Mycoplasma orale and Mycoplasma hyorhinis, as well as less common species
  • Addresses widespread contamination challenges that can result in significant industry losses
  • Resistant to antibiotics targeting cell wall formation, such as penicillin
  • Lack of cell wall enables passage through 0.2 μm filters
  • Prevalence in cell cultures reported between 25% and 80%
  • Major contamination sources include human, porcine, and bovine hosts
  • Detects multiple species including Acholeplasma laidlawii, Mycoplasma arginine, Mycoplasma fermentans, Mycoplasma hominis, Mycoplasma hyorhinis, Mycoplasma orale, Mycoplasma salivarium, and Mycoplasma pneumoniae
  • Highly sensitive, specific, and rapid assay targeting conserved Mollicutes genome regions
  • No cross-reactivity with bacterial, fungal, or mammalian DNA
  • Simple to perform without the need for special technical expertise or lab equipment
  • Positive results visualized on an immunochromatographic test device in under 5 minutes after loading
  • Application notes highlight use for simple, sensitive, rapid detection of mycoplasma in cell cultures
  • Key applications include mycoplasma detection and cell culture research
  • Research Use Only (RUO)

Spezifikation

Form Liquid
Product Type Mycoplasma Detection
Sterility Non-sterile
Quantity 5 Tests
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.