missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ Mycoalert™ Mycoplasma Detection Kit

Product Code. 10255393
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
50 Tests
Unit Size:
1 set
25 tests
50 tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10255393 100 Tests 1 set
11650261 25 Tests 25 tests
11630271 50 Tests 50 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10255393 Supplier Lonza™ Supplier No. LT07318
  • ONLINE EXCLUSIVE PRICE
    2010.00 €
    1718.04 € / 1 sets
    Save 291.96 €
    14% Off
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect mycoplasma in culture. Mycoalert™ Mycoplasma Detection Kit is a selective biochemical test that exploits the activity of mycoplasmal enzymes which are found in all six of the main mycoplasma cell culture contaminants and the vast majority of 180 mycoplasma species, but are not present in eukaryotic cells.

Convenient & fast
  • Simply add two reagents to the culture supernatant
  • Perform two luminescence readings
  • Results in < 20 min
  • Easy interpretation of results
Universal
  • Detects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma; except Ureaplasma)
  • Negligible interference with media components
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Mycoalert reagent: 1 x 10mL, Mycoalert assay buffer: 1 x 20mL, Mycoalert substrate: 1 x 10mL
2 to 8°C
Cell Type Mammalian Cells
Product Type Mycoplasma Detection
Quantity 100 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.