missing translation for 'onlineSavingsMsg'
Learn More

Mycophenolate mofetil, 98%

Product Code. 15579583 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15579583 1 g 1g
15589583 5 g 5g
15599583 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15579583 Supplier Thermo Scientific Acros Supplier No. 459370010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 128794-94-5 | C23H31NO7 | 433.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128794-94-5
Molecular Formula C23H31NO7
Molecular Weight (g/mol) 433.5
InChI Key RTGDFNSFWBGLEC-SYZQJQIISA-N
Synonym mycophenolate mofetil, cellcept, munoloc, myfenax, mycophenolic acid morpholinoethyl ester, tm-mmf, mycophenylate mofetil, mycophenolatemofetil, cellcept tn, unii-9242ecw6r0
PubChem CID 5281078
ChEBI CHEBI:8764
IUPAC Name 2-morpholin-4-ylethyl (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoate
SMILES CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)OCCN3CCOCC3)OC

Specifications

Melting Point 95.0°C to 96.0°C
Ignition Residue 0.1% max
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 433.5
Percent Purity 98%
Chemical Name or Material Mycophenolate mofetil
compliance-icons compliance-icons
Product Identifier
  • Mycophenolate mofetil
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to an approved waste d.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.